Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobenzophenone 97.0+%, TCI America™
SDP

Catalog No. B39371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39371G 1 g
B39375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39371G Supplier TCI America Supplier No. B39371G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13047-06-8
Molecular Formula C13H9BrO
Molecular Weight (g/mol) 261.12
MDL Number MFCD00032462
InChI Key ABEVIHIQUUXDMS-UHFFFAOYSA-N
PubChem CID 2735534
IUPAC Name (2-bromophenyl)(phenyl)methanone
SMILES BrC1=CC=CC=C1C(=O)C1=CC=CC=C1

Specifications

Melting Point 42°C
Color Yellow
Boiling Point 163°C
Quantity 1 g
Formula Weight 261.12
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromobenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.