Learn More
2-Bromobutane, 98%
Description
2-Bromobutane is utilized in the synthesis of non-nucleosides. It is used as a solvent as well as an intermediate in organic synthesis. It is an important raw material used to prepare Grignard reagent by reacting with magnesium. Further, it reacts with hydrocyanic acid to prepare sodium salt to get (+-)-sec-butyl cyanide. It is an important raw material used to prepare Grignard reagent by reacting with magnesium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Bromobutane is utilized in the synthesis of non-nucleosides. It is used as a solvent as well as an intermediate in organic synthesis. It is an important raw material used to prepare Grignard reagent by reacting with magnesium. Further, it reacts with hydrocyanic acid to prepare sodium salt to get (+-)-sec-butyl cyanide. It is an important raw material used to prepare Grignard reagent by reacting with magnesium.
Solubility
Miscible with alcohol and ether. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, strong bases, magnesium, sodium, sodium oxides and potassium.
Specifications
Specifications
| Melting Point | °C to 112°C |
| Density | 1.260 |
| Boiling Point | 91°C to 92°C |
| Flash Point | 13°C (55°F) |
| Assay Percent Range | 98% |
| Refractive Index | 1.4369 |
| Linear Formula | CH3CHBrCH2CH3 |
| Quantity | 250 g |
| UN Number | UN2339 |
| Beilstein | 505949 |
| Show More |
Safety and Handling
| Hazard Category | H225-H315-H319-H335 |
| Hazard Statement | GHS H Statement H225-H336 Highly flammable liquid and vapour. May cause drowsiness or dizziness. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-BROMOBUTANE |
| EINECSNumber | 201-140-7 |
| RTECSNumber | EJ6228000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.