Learn More
2-Bromodibenzofuran, 98%, Thermo Scientific Chemicals
Description
2-Bromodibenzofuran is used as a reagent in Friedel-Crafts acylation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Bromodibenzofuran is used as a reagent in Friedel-Crafts acylation reactions.
Solubility
Soluble in hot methanol.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 108°C to 111°C |
| Quantity | 5 g |
| Beilstein | 142199 |
| Solubility Information | Soluble in hot methanol. |
| Formula Weight | 247.09 |
| Percent Purity | 98% |
| Physical Form | Solid |
| Chemical Name or Material | 2-Bromodibenzofuran |
Safety and Handling
| Hazard Category | H302-H319 |
| Hazard Statement | GHS H Statement H302-H319 Harmful if swallowed. Causes serious eye irritation. |
| Precautionary Statement | P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.