Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromoethyl methyl ether

Catalog No. 23815525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-815-525G 25 g
23-815-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-815-525G Supplier Sigma Aldrich Supplier No. 23815525G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Kinetics of potassium iodide exchange reaction with 2-bromoethyl methyl ether in acetone at 15 and 25C has been investigated.2-Bromoethyl methyl ether has been used as starting reagent in the synthesis of 1-bromo-4-(2-methoxyethoxy)benzene and 1-bromo-4-[(2-methoxyethoxy)methyl]benzene.

Specifications

CAS 6482-24-2
Density 1.479 g/mL (at 25°C (literature))
Boiling Point 40°C to 41°C (66 mmHg)
Molecular Formula C3H7BrO
Linear Formula BrCH2CH2OCH3
Refractive Index n20/D 1.447 (literature)
MDL Number MFCD00000236
Quantity 25 g
Synonym 1-Bromo-2-methoxyethane
Molecular Weight (g/mol) 138.99
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.