Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromoethyl methyl ether

Catalog No. 2381555G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-815-55G 5 g
23-815-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-815-55G Supplier Sigma Aldrich Supplier No. 2381555G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Kinetics of potassium iodide exchange reaction with 2-bromoethyl methyl ether in acetone at 15 and 25C has been investigated.2-Bromoethyl methyl ether has been used as starting reagent in the synthesis of 1-bromo-4-(2-methoxyethoxy)benzene and 1-bromo-4-[(2-methoxyethoxy)methyl]benzene.

Specifications

CAS 6482-24-2
Density 1.479 g/mL (at 25°C (literature))
Boiling Point 40°C to 41°C (66 mmHg)
Molecular Formula C3H7BrO
Refractive Index n20/D 1.447 (literature)
Linear Formula BrCH2CH2OCH3
MDL Number MFCD00000236
Quantity 5 g
Synonym 1-Bromo-2-methoxyethane
Molecular Weight (g/mol) 138.99
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.