Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromofluorene, 95%

Catalog No. AC154220050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC154220050 5 g
AC154220250 25 g
AC154221000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC154220050 Supplier Thermo Scientific Chemicals Supplier No. 154220050

CAS: 1133-80-8 | C13H9Br | 245.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1133-80-8
MDL Number MFCD00001115
InChI Key FXSCJZNMWILAJO-UHFFFAOYSA-N
Synonym 2-bromofluorene, 9h-fluorene, 2-bromo, 2-brom-9h-fluoren, 2-bromofluorene, tech., #, acmc-1c7b2, ksc181a7p, 2-bromofluorene 5g, 4-bromo-2,2'-methylenebiphenyl
PubChem CID 14336
IUPAC Name 2-bromo-9H-fluorene
SMILES C1C2=CC=CC=C2C3=C1C=C(C=C3)Br

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 110°C to 115°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C13H9Br
Quantity 5 g
Beilstein 05, II, 534
Molecular Weight (g/mol) 245.119
Formula Weight 245.12
Percent Purity ≥94%
Physical Form Slightly Crystalline Powder or Flakes
Chemical Name or Material 2-Bromofluorene
Show More Show Less
missing translation for 'healthHazard2' GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.