Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromofluorene 98.0+%, TCI America™
SDP

Catalog No. B306925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B306925G 25 g
B30695G 5 g
B3069100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B306925G Supplier TCI America Supplier No. B306925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1133-80-8
Molecular Formula C13H9Br
Molecular Weight (g/mol) 245.119
MDL Number MFCD00001115
InChI Key FXSCJZNMWILAJO-UHFFFAOYSA-N
Synonym 2-bromofluorene, 9h-fluorene, 2-bromo, 2-brom-9h-fluoren, 2-bromofluorene, tech., #, acmc-1c7b2, ksc181a7p, 2-bromofluorene 5g, 4-bromo-2,2'-methylenebiphenyl
PubChem CID 14336
IUPAC Name 2-bromo-9H-fluorene
SMILES C1C2=CC=CC=C2C3=C1C=C(C=C3)Br

Specifications

Melting Point 114°C
Color White-Yellow
Boiling Point 185°C
Quantity 25 g
Formula Weight 245.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromofluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.