Learn More
Selleck Chemical LLC 2-BP-E0120-5MG
SDP
Supplier:  Selleck Chemical LLC E01205MG
Encompass_Preferred
2-BP is a nonmetabolisable analogue of palmitate and acts as a palmitoylation inhibitor which inhibits multiple palmitoyl-acyl transferase (PAT) activity towards MyrGCK(NBD) and FarnCNRas(NBD) with identical IC50s of approx 4 M 2-Bromohexadecanoic acid can directly and irreversibly inhibit the palmitoyltransferase activity of all DHHC (Asp-His-His-Cys) proteins
Catalog No. 50-234-1089
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.