Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoindene 98.0+%, TCI America™
SDP

Catalog No. p-7119993 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B31635G Supplier TCI America Supplier No. B31635G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10485-09-3
Molecular Formula C9H7Br
Molecular Weight (g/mol) 195.059
MDL Number MFCD06797863
InChI Key CCUYEVNCRQDQRF-UHFFFAOYSA-N
Synonym 2-bromoindene, 1h-indene, 2-bromo, indene, 2-bromo, 2-bromanyl-1h-indene, 2-bromoindene, 95+%, 2-bromo-indene, pubchem9657, #, acmc-1c5rn, sodium perborate,tetrahydrate
PubChem CID 575586
IUPAC Name 2-bromo-1H-indene
SMILES C1C2=CC=CC=C2C=C1Br

Specifications

Melting Point 38°C
Boiling Point 127°C
Quantity 5 g
Formula Weight 195.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoindene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.