Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoisobutyryl Bromide 98.0+%, TCI America™
SDP

Catalog No. B0607100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0607100G 100 g
B060725G 25 g
B0607500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0607100G Supplier TCI America Supplier No. B0607100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20769-85-1
Molecular Formula C4H6Br2O
Molecular Weight (g/mol) 229.899
MDL Number MFCD00000122
InChI Key YOCIJWAHRAJQFT-UHFFFAOYSA-N
Synonym 2-bromo-2-methylpropionyl bromide, 2-bromoisobutyryl bromide, propanoyl bromide, 2-bromo-2-methyl, bibb, 2-bromo-2-methylpropionylbromide, unii-627f9n9dya, 2-bromoisobutyrylbromide, 2-bromo-2-methyl-propionyl bromide, alpha-bromoisobutyryl bromide, 2-bromo-2-methyl-propanoyl bromide
PubChem CID 88685
IUPAC Name 2-bromo-2-methylpropanoyl bromide
SMILES CC(C)(C(=O)Br)Br

Specifications

Color Yellow
Boiling Point 164°C
Quantity 100 g
UN Number 3265
Formula Weight 229.90
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromoisobutyryl Bromide
EINECSNumber (2)-3443
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.