Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromomethyl-4-fluorobenzonitrile, 98%

Catalog No. AAH6311006
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6311006 5 g
AAH6311014 25 g
AAH6311003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6311006 Supplier Thermo Scientific Chemicals Supplier No. H6311006
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 421552-12-7 | C8H5BrFN | 214.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 421552-12-7
Molecular Formula C8H5BrFN
Molecular Weight (g/mol) 214.04
MDL Number MFCD08059542
InChI Key CHCAGFNTASDQFX-UHFFFAOYSA-N
Synonym 2-cyano-5-fluorobenzyl bromide, 2-bromomethyl-4-fluorobenzonitrile, 2-cyano-5-fluorobenzylbromide, 2-cyano-5-fluoro benzyl bromide, 2-bromomethyl-4-fluoro-benzonitrile, 2-bromomethyl-4-fluorobenzenecarbonitrile, pubchem4906, acmc-1arut, ksc493s9t, 2-cyano-5-flurobenzyl bromide
PubChem CID 21984990
IUPAC Name 2-(bromomethyl)-4-fluorobenzonitrile
SMILES FC1=CC(CBr)=C(C=C1)C#N

Specifications

Quantity 5 g
UN Number UN2923
Formula Weight 214.04
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromomethyl-4-fluorobenzonitrile
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.