Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromophenacyl Bromide 97.0+%, TCI America™
SDP

Catalog No. B41241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41241G 1 g
B41245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41241G Supplier TCI America Supplier No. B41241G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 49851-55-0
Molecular Formula C8H6Br2O
Molecular Weight (g/mol) 277.943
MDL Number MFCD00044660
InChI Key LAXPJIJQTHJGCK-UHFFFAOYSA-N
PubChem CID 2737522
IUPAC Name 2-bromo-1-(2-bromophenyl)ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)Br

Specifications

Color Yellow
Boiling Point 100°C
Quantity 1 g
UN Number 3265
Synonym 2,2′C-Dibromoacetophenone
Formula Weight 277.94
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromophenacyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.