Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromophenanthrene 97.0+%, TCI America™
SDP

Catalog No. B46711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46711G 1 g
B4671200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46711G Supplier TCI America Supplier No. B46711G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62162-97-4
Molecular Formula C14H9Br
Molecular Weight (g/mol) 257.13
InChI Key SQTPFYJEKHTINP-UHFFFAOYSA-N
PubChem CID 12491433
IUPAC Name 2-bromophenanthrene
SMILES C1=CC=C2C(=C1)C=CC3=C2C=CC(=C3)Br

Specifications

Melting Point 97°C
Color White-Yellow
Boiling Point 185°C
Quantity 1 g
Formula Weight 257.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromophenanthrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.