Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Bromophenyl)diphenylphosphine 95.0+%, TCI America™
SDP

Catalog No. B39531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39531G 1 g
B39535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39531G Supplier TCI America Supplier No. B39531G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62336-24-7
Molecular Formula C18H14BrP
Molecular Weight (g/mol) 341.19
MDL Number MFCD00045014
InChI Key XIONUQPOXCUMMB-UHFFFAOYSA-N
PubChem CID 112836
IUPAC Name (2-bromophenyl)diphenylphosphane
SMILES BrC1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 112°C
Color White
Quantity 1 g
Formula Weight 341.19
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2-Bromophenyl)diphenylphosphine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.