Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromophenyl p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. B45281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45281G 1 g
B45285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45281G Supplier TCI America Supplier No. B45281G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84672-48-0
Molecular Formula C13H11BrO3S
Molecular Weight (g/mol) 327.192
MDL Number MFCD19707445
InChI Key ZXJVRRGBOXUFSQ-UHFFFAOYSA-N
Synonym p-Toluenesulfonic Acid 2-Bromophenyl Ester, 2-Bromophenyl Tosylate
PubChem CID 262269
IUPAC Name (2-bromophenyl) 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC2=CC=CC=C2Br

Specifications

Melting Point 76°C
Color White
Quantity 1 g
Formula Weight 327.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromophenyl p-Toluenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.