Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromopropionyl Chloride 97.0+%, TCI America™
SDP

Catalog No. B4107100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4107100G 100 g
B410725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4107100G Supplier TCI America Supplier No. B4107100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7148-74-5
Molecular Formula C3H4BrClO
Molecular Weight (g/mol) 171.42
MDL Number MFCD00000711
InChI Key OZGMODDEIHYPRY-UHFFFAOYNA-N
Synonym 2-bromopropionyl chloride, propanoyl chloride, 2-bromo, 2-bromopropionylchloride, 2-bromo-propionyl chloride, 2-brominepropionylchloride, alpha-bromopropionyl chloride, 2-bromopropionic acid chloride, sgqdjlpqbsmuh@, 2-bromopropanoylchloride, acmc-1cjhb
PubChem CID 97980
IUPAC Name 2-bromopropanoyl chloride
SMILES CC(Br)C(Cl)=O

Specifications

Color Yellow
Boiling Point 131°C
Quantity 100 g
UN Number 2920
Formula Weight 171.42
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Bromopropionyl Chloride
EINECSNumber (9)-1148&(9)-1184&(9)-1234
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.