Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromopyridine 98.0+%, TCI America™
SDP

Catalog No. B0650100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0650100ML 100 mL
B065025ML 25 mL
B0650500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0650100ML Supplier TCI America Supplier No. B0650100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 109-04-6
Molecular Formula C5H4BrN
Molecular Weight (g/mol) 157.998
MDL Number MFCD00006219
InChI Key IMRWILPUOVGIMU-UHFFFAOYSA-N
Synonym o-bromopyridine, pyridine, 2-bromo, 2-bromo-pyridine, 2-pyridyl bromide, alpha-bromopyridine, beta-bromopyridine, bromopyridine, 2-bromopyridin, 2-bromo pyridine, unii-7z7mlc4vd8
PubChem CID 7973
ChEBI CHEBI:51574
IUPAC Name 2-bromopyridine
SMILES C1=CC=NC(=C1)Br

Specifications

Color Yellow
Boiling Point 194°C
Quantity 100 mL
UN Number 2929
Formula Weight 158.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromopyridine
RTECSNumber US3850000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.