Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoquinoxaline 98.0+%, TCI America™
SDP

Catalog No. B46321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46321G 1 g
B4632200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46321G Supplier TCI America Supplier No. B46321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 36856-91-4
Molecular Formula C8H5BrN2
Molecular Weight (g/mol) 209.046
MDL Number MFCD07368206
InChI Key XVKLMYQMVPHUPN-UHFFFAOYSA-N
Synonym quinoxaline, 2-bromo, bromoquinoxaline, 2-bromoquinoxaline, ksc226a0j
PubChem CID 582225
IUPAC Name 2-bromoquinoxaline
SMILES C1=CC=C2C(=C1)N=CC(=N2)Br

Specifications

Melting Point 57°C
Quantity 1 g
Formula Weight 209.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoquinoxaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.