Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromothiophene, 98%

Catalog No. p-4510269
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154290500 50 mL Glass bottle
AC154292500 250 mL Glass bottle
AC154290010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC154290500 Supplier Thermo Scientific Chemicals Supplier No. 154290500
Only null left
Add to Cart

CAS: 1003-09-4 | C4H3BrS | 163.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-09-4
Molecular Formula C4H3BrS
Molecular Weight (g/mol) 163.03
MDL Number MFCD00005417
InChI Key TUCRZHGAIRVWTI-UHFFFAOYSA-N
Synonym 2-thienyl bromide, thiophene, 2-bromo, thiophene, bromo, bromothiophene, 2-bromo-thiophene, 2-bromo thiophene, thienyl bromide, 5-bromothiophene, 5-bromo-thiophene, 2-bromanylthiophene
PubChem CID 13851
IUPAC Name 2-bromothiophene
SMILES BrC1=CC=CS1

Specifications

Melting Point -10.0°C
Color Brown to Colorless
Density 1.6800g/mL
Boiling Point 149.0°C to 151.0°C
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5850 to 1.5870
Quantity 50 mL
Beilstein 17,33
Specific Gravity 1.68
Solubility Information Solubility in water: immiscible. Other solubilities: very soluble in ether, acetone, soluble in carbontetrachloride
Formula Weight 163.04
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromothiophene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses
EINECSNumber 213-699-4
Recommended Storage Product darkens on exposure to light

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.