Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butanol, puriss. p.a., Reag. Ph. Eur., ≥99.5% (GC), Solstice

Catalog No. 60045074 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-074 1 L Glass Bottle
60-045-075 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-074 Supplier Solstice Supplier No. 194401L
Add to Cart
Add to Cart

Puriss. p.a., Reag. Ph. Eur.,≥99.5% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-92-2
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004569
InChI Key BTANRVKWQNVYAZ-UHFFFAOYNA-N
Synonym 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
PubChem CID 6568
ChEBI CHEBI:35687
IUPAC Name butan-2-ol
SMILES CCC(C)O

Specifications

Melting Point -115°C
Color Colorless
Density 0.808g/mL at 25°C
Quantity 1 L
Explosion Limit 0.098
Flash Point 24°C (80.6°F)
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.397
Linear Formula CH3CH2CH(OH)CH3
Vapor Pressure 12.5mmHg (20°C)
UN Number UN1120
Beilstein 773649
Boiling Point 99°C
Formula Weight 74.12g/mol
Percent Purity ≥99.5% (GC)
Grade puriss. p.a.
Chemical Name or Material 2-Butanol, Reag. Ph. Eur.
Show More Show Less
Hazard Statement P210-P305 + P351 + P338-P370 + P378
Precautionary Statement H226-H319-H335-H336
DOTInformation Class 3 - PG 3 - Butanols
EINECSNumber 201-158-5
RTECSNumber EO1750000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.