Learn More
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Melting Point | -87°C (literature) |
| CAS Max % | 100.0000% |
| Density | 0.805 g/mL at 25 °C(lit.) |
| Quantity | 2.5 L |
| Flash Point | −4°C |
| Packaging | Glass Bottle |
| Linear Formula | C2H5COCH3 |
| UN Number | UN1193 |
| Beilstein | 741880 |
| Synonym | 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon |
| Show More |
Safety and Handling
| ShelfLife | 1260 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl methyl ketone |
| EINECSNumber | 201-159-0 |
| Recommended Storage | Room Temp |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.