Learn More
2-Butanone, Reagent, ACS, 99%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical 2-Butanone, Reagent, ACS
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 6 x 1 L |
| Packaging | Amber Glass Bottle |
| Residue after Evaporation | 0.0025% |
| Titratable Acid | 0.0005 meq/g |
| Water | 0.2% |
| UN Number | UN1193 |
| Boiling Point | 80 °C |
| Percent Purity | 99% |
| Grade | Reagent |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.