Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butoxyethanol, (Ethylene Glycol Monobutyl Ether), Reagents

Catalog No. 01337383
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-383 20 L
01-337-384 1 L
01-337-382 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-383 Supplier Reagents Holdings Llc Supplier No. C215625020E6
Add to Cart
Add to Cart

2-Butoxyethanol, (Ethylene Glycol Monobutyl Ether), NIST Traceable, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 111-76-2
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
InChI Key POAOYUHQDCAZBD-UHFFFAOYSA-N
Synonym Ethylene Glycol Monobutyl Ether, Butyl Cellosolve
IUPAC Name 2-butoxyethan-1-ol
SMILES CCCCOCCO

Specifications

Boiling Range 171°C
Melting Point -70°C
Color Colorless
Boiling Point 171°C
Melting Range -70°C
Packaging Steel Pail
Quantity 20 L
Specific Gravity 0.9019
Physical Form Liquid
Chemical Name or Material 2-Butoxyethanol, (Ethylene Glycol Monobutyl Ether)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.