Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Butoxyethyl acetate

Catalog No. 30728925ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-728-925ML 25 mL
30-728-91L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-728-925ML Supplier Sigma Aldrich Supplier No. 30728925ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-butoxyethyl acetate is one of the components of jiashi melon juice and was identified by gas chromatography-mass spectrometry-olfactometry (GC-ms-o) analysis
  • It stimulated the release of prostaglandin e(2), an arachidonic acid metabolite, in human epidermal keratinocytes

Specifications

CAS 112-07-2
Density 0.942 g/mL (at 25°C (literature))
Boiling Point 192°C (lit.)
Molecular Formula C8H16O3
Refractive Index n20/D 1.413 (literature)
Linear Formula CH3COOCH2CH2O(CH2)3 CH3
Quantity 25 mL
MDL Number MFCD00009457
Synonym 1-Acetoxy-2-butoxyethane; BCA; Ethylene glycol monobutyl ether acetate
Molecular Weight (g/mol) 160.21
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.