Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butoxyethyl acetate, 98%

Catalog No. AC403410025
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC403410025 2.5 L
AC403410010 1 L
AC403410100 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC403410025 Supplier Thermo Scientific Chemicals Supplier No. 403410025
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 112-07-2 | C8H16O3 | 160.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-07-2
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.21
MDL Number MFCD00009457
InChI Key NQBXSWAWVZHKBZ-UHFFFAOYSA-N
Synonym butoxyethyl acetate, butyl glycol acetate, butylglycol acetate, ethylene glycol monobutyl ether acetate, 2-butoxyethanol acetate, butyl cellosolve acetate, ektasolve eb acetate, ethanol, 2-butoxy-, acetate, butylcelosolvacetat, 1-acetoxy-2-butoxyethane
PubChem CID 8160
IUPAC Name 2-butoxyethyl acetate
SMILES CCCCOCCOC(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -64.0°C
Density 0.9385g/mL
Boiling Point 193.5°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4120 to 1.4140
Linear Formula CH3CO2CH2CH2O(CH2)3CH3
Quantity 2.5 L
Specific Gravity 0.9385
Solubility Information Solubility in water: soluble
Formula Weight 160.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Butoxyethyl acetate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 203-933-3
RTECSNumber KJ8925000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.