Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butyl-1H-imidazole-5-carboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. B406425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B406425G 25 g
B40645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B406425G Supplier TCI America Supplier No. B406425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68282-49-5
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.197
MDL Number MFCD01075030
InChI Key PTHGVOCFAZSNNA-UHFFFAOYSA-N
Synonym 2-butyl-1h-imidazole-4-carbaldehyde, 2-butyl-5-formylimidazole, 2-butyl-4-formylimidazole, 2-butyl-4-imidazolyl formaldehyde, unii-u12g73sqsx, 2-butyl-1h-imidazole-5-carboxaldehyde, u12g73sqsx, 1h-imidazole-5-carboxaldehyde, 2-butyl, 2-butylimidazole-4-carbaldehyde, 2-butyl-3h-imidazole-4-carbaldehyde
PubChem CID 2735671
IUPAC Name 2-butyl-1H-imidazole-5-carbaldehyde
SMILES CCCCC1=NC=C(N1)C=O

Specifications

Melting Point 118°C
Color Yellow
Quantity 25 g
Formula Weight 152.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Butyl-1H-imidazole-5-carboxaldehyde
EINECSNumber (5)-6676
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.