Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butyl-n-octan-1-amine 98.0+%, TCI America™
SDP

Catalog No. B55501ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55501ML 1 mL
B55505ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55501ML Supplier TCI America Supplier No. B55501ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 217655-07-7
Molecular Formula C12H27N
Synonym 2-Butyl-1-n-octylamine, 1-Amino-2-butyl-n-octane, 5-(Aminomethyl)undecane

Specifications

Color Colorless
Quantity 1 mL
UN Number 2735
Formula Weight 185.36
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Butyl-n-octan-1-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.