Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butyn-1-ol 98.0+%, TCI America™
SDP

Catalog No. B071225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B071225ML 25 mL
B07125ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B071225ML Supplier TCI America Supplier No. B071225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 764-01-2
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00002914
InChI Key NEEDEQSZOUAJMU-UHFFFAOYSA-N
Synonym 2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol
PubChem CID 12991
IUPAC Name but-2-yn-1-ol
SMILES CC#CCO

Specifications

Color Yellow
Boiling Point 145°C
Quantity 25 mL
UN Number 1987
Formula Weight 70.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Butyn-1-ol
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.