Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butynyl p-toluenesulfonate, 98%, Thermo Scientific™

Catalog No. AAL1249906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1249906 5 g
AAL1249903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1249906 Supplier Thermo Scientific Chemicals Supplier No. L1249906

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56563-37-2
Molecular Formula C11H12O3S
Molecular Weight (g/mol) 224.274
MDL Number MFCD00078395
InChI Key HGZKJNHJABSJTC-UHFFFAOYSA-N
Synonym 2-butynyl 4-toluenesulfonate, 2-butynyl 4-methylbenzenesulfonate, but-2-yn-1-yl 4-methylbenzenesulfonate, 2-butynyl p-toluenesulfonate, acmc-1ax7n, 2-butynyl4-toluenesulfonate, p-toluenesulfonic acid 2-butynyl ester, p-toluenesulphonic acid-2-butinyl ester, but-2-yn-1-yl 4-methylbenzene-1-sulfonate
PubChem CID 7018873
IUPAC Name but-2-ynyl 4-methylbenzenesulfonate
SMILES CC#CCOS(=O)(=O)C1=CC=C(C=C1)C

Specifications

Melting Point 47°C to 48°C
Quantity 5 g
Beilstein 3300477
Formula Weight 224.28
Percent Purity 98%
Chemical Name or Material 2-Butynyl p-toluenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.