Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Carbobenzoxyamino)-2-hydroxyacetic Acid 98.0+%, TCI America™
SDP

Catalog No. C26811G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26811G 1 g
C26815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26811G Supplier TCI America Supplier No. C26811G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56538-57-9
Molecular Formula C10H11NO5
Molecular Weight (g/mol) 225.2
MDL Number MFCD02179279
InChI Key JQEZLSUFDXSIEK-UHFFFAOYSA-N
Synonym 2-(Cbz-amino)-2-hydroxyacetic Acid, N-Carbobenzoxy-2-hydroxyglycine, Z-2-hydroxyglycine
PubChem CID 3509506
IUPAC Name 2-hydroxy-2-(phenylmethoxycarbonylamino)acetic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(C(=O)O)O

Specifications

Melting Point 189°C
Color White
Quantity 1 g
Formula Weight 225.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Carbobenzoxyamino)-2-hydroxyacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.