Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Carbobenzoxyamino)isobutyric Acid 98.0+%, TCI America™
SDP

Catalog No. C276125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C276125G 25 g
C27615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C276125G Supplier TCI America Supplier No. C276125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15030-72-5
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00004190
InChI Key QKVCSJBBYNYZNM-UHFFFAOYSA-N
Synonym z-aib-oh, n-cbz-2-methylalanine, 2-benzyloxy carbonyl amino-2-methylpropanoic acid, n-cbz-2-amino-2-methylpropanoic acid, z-2-aminoisobutyric acid, z-2-methylalanine, n-carbobenzyloxy-2-methylalanine, 2-carbobenzoxyamino isobutyric acid, n-benzyloxy carbonyl-2-methylalanine, alanine, 2-methyl-n-phenylmethoxy carbonyl
PubChem CID 294936
IUPAC Name 2-methyl-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES CC(C)(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 79°C
Color White
Quantity 25 g
Formula Weight 237.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Carbobenzoxyamino)isobutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.