Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1-(4-fluorobenzyl)benzimidazole 98.0+%, TCI America™
SDP

Catalog No. C285025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C285025G 25 g
C28505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C285025G Supplier TCI America Supplier No. C285025G

Chemical Identifiers

CAS 84946-20-3
Molecular Formula C14H10ClFN2
Molecular Weight (g/mol) 260.696
MDL Number MFCD00800240
InChI Key PGXALMVNIRPELS-UHFFFAOYSA-N
PubChem CID 3020241
IUPAC Name 2-chloro-1-[(4-fluorophenyl)methyl]benzimidazole
SMILES C1=CC=C2C(=C1)N=C(N2CC3=CC=C(C=C3)F)Cl

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 25 g
Formula Weight 260.70
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-1-(4-fluorobenzyl)benzimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.