Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1,4-phenylenediamine Sulfate 98.0+%, TCI America™
SDP

Catalog No. C025625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C025625G 25 g
C0256500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C025625G Supplier TCI America Supplier No. C025625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61702-44-1
Molecular Formula C6H9ClN2O4S
Molecular Weight (g/mol) 240.66
MDL Number MFCD00013004
InChI Key GQFGHCRXPLROOF-UHFFFAOYSA-N
Synonym 2-chloro-p-phenylenediamine sulfate, 2-chlorobenzene-1,4-diamine sulfate, 2-chloro-1,4-phenylenediamine sulfate, fourrine so, 2-chloro-1,4-benzenediamine sulfate, 2-chlorobenzene-1,4-diamine; sulfuric acid, 1,4-benzenediamine, 2-chloro-, sulfate, renal so, fourrine 81, 2-cl-p-pd
PubChem CID 22584
ChEBI CHEBI:76597
IUPAC Name 2-chlorobenzene-1,4-diamine; sulfuric acid
SMILES OS(O)(=O)=O.NC1=CC=C(N)C(Cl)=C1

Specifications

Melting Point 253°C
Color White-Yellow
Quantity 25 g
Formula Weight 240.66
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-1,4-phenylenediamine Sulfate
RTECSNumber CZ1581000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.