Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-2',4'-difluoroacetophenone 98.0+%, TCI America™
SDP

Catalog No. C203025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C203025G 25 g
C20305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C203025G Supplier TCI America Supplier No. C203025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51336-94-8
Molecular Formula C8H5ClF2O
Molecular Weight (g/mol) 190.574
InChI Key UENGBOCGGKLVJJ-UHFFFAOYSA-N
Synonym 2-chloro-2',4'-difluoroacetophenone, 2-chloro-1-2,4-difluorophenyl ethanone, 2,4-difluorophenacyl chloride, 2-chloro-2,4-difluoroacetophenone, 2',4'-difluoro-alfa-chloroacetophenone, 2-chloro-1-2,4-difluorophenyl ethan-1-one, 2',4'-difluoro-2-chloroacetophenone, 1-2,4-difluorophenyl-2-chloroethan-1-one, 2',4'-difluorophenacyl chloride, 2'-chloro-2,4-difluoroacetophenone
PubChem CID 588083
IUPAC Name 2-chloro-1-(2,4-difluorophenyl)ethanone
SMILES C1=CC(=C(C=C1F)F)C(=O)CCl

Specifications

Melting Point 50°C
Quantity 25 g
UN Number 3261
Formula Weight 190.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-2′,4′-difluoroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.