Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-(chloromethyl)quinoline 98.0+%, TCI America™
SDP

Catalog No. C28611G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28611G 1 g
C28615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28611G Supplier TCI America Supplier No. C28611G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90097-52-2
Molecular Formula C10H7Cl2N
Molecular Weight (g/mol) 212.073
MDL Number MFCD05865134
InChI Key UOIJHUIOTFDGNA-UHFFFAOYSA-N
Synonym 2-chloro-3-chloromethyl quinoline, quinoline, 2-chloro-3-chloromethyl, 2-chloranyl-3-chloromethyl quinoline
PubChem CID 2063384
IUPAC Name 2-chloro-3-(chloromethyl)quinoline
SMILES C1=CC=C2C(=C1)C=C(C(=N2)Cl)CCl

Specifications

Melting Point 117°C
Color Yellow
Quantity 1 g
UN Number 3261
Formula Weight 212.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-(chloromethyl)quinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.