Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-methoxybenzoic acid, 97%

Catalog No. AC459900050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459900050 5 g
AC459900010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459900050 Supplier Thermo Scientific Chemicals Supplier No. 459900050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 33234-36-5 | C8H7ClO3 | 186.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33234-36-5
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.59
MDL Number MFCD07784399
InChI Key VIUDWLLKFANPLX-UHFFFAOYSA-N
Synonym benzoic acid, 2-chloro-3-methoxy, acmc-20a6cl, 2-chloro-m-anisic acid, 2-chloro-3-methoxybenzoicacid, 2-chloro-3-methoxybenzoic acid
PubChem CID 16218063
IUPAC Name 2-chloro-3-methoxybenzoic acid
SMILES COC1=CC=CC(C(O)=O)=C1Cl

Specifications

Melting Point 161.0°C to 165.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 186.6
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-methoxybenzoic acid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed. Causes serious eye irritation.
Precautionary Statement GHS P Statement Wear eye protection/face protection. Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.