Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-nitrotoluene 98.0+%, TCI America™
SDP

Catalog No. C26111G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C26111G Supplier TCI America Supplier No. C26111G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3970-40-9
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00134653
InChI Key XTSGZXRUCAWXKY-UHFFFAOYSA-N
Synonym 2-chloro-3-nitrotoluene, 2-chloro-3-methylnitrobenzene, 2-chloro-3-nitro toluene, benzene, 2-chloro-1-methyl-3-nitro, chloronitrotoluene, ksc493s7b, 2-?chloro-?3-?nitrotoluene
PubChem CID 77591
IUPAC Name 2-chloro-1-methyl-3-nitrobenzene
SMILES CC1=CC=CC(=C1Cl)[N+]([O-])=O

Specifications

Color Yellow
Boiling Point 148°C
Quantity 1 g
UN Number 2433
Formula Weight 171.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-3-nitrotoluene
EINECSNumber (3)-0449
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.