Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4,5-difluorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. C1460250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1460250G 250 g
C146025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1460250G Supplier TCI America Supplier No. C1460250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 110877-64-0
Molecular Formula C7H2ClF2O2
Molecular Weight (g/mol) 191.54
MDL Number MFCD00077479
InChI Key CGFMLBSNHNWJAW-UHFFFAOYSA-M
Synonym 2-chloro-4,5-difluoro-benzoic acid, 2'-chloro-4',5'-difluorobenzoic acid, benzoic acid, 2-chloro-4,5-difluoro, pubchem1339, ksc177o4l, rarechem al bo 1000, timtec-bb sbb003616, 2-chloro-4,5-difluorobenzoic acid, benzoicacid, 2-chloro-4,5-difluoro
PubChem CID 737171
IUPAC Name 2-chloro-4,5-difluorobenzoate
SMILES [O-]C(=O)C1=CC(F)=C(F)C=C1Cl

Specifications

Melting Point 105°C
Color White
Quantity 250 g
Formula Weight 192.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4,5-difluorobenzoic Acid
EINECSNumber (3)-4165
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.