Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-ethoxybenzeneboronic acid, 95%, Thermo Scientific™

Catalog No. AAH5299003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5299003 1 g
AAH52990MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5299003 Supplier Thermo Scientific Chemicals Supplier No. H5299003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313545-44-7
Molecular Formula C8H10BClO3
Molecular Weight (g/mol) 200.43
MDL Number MFCD02684318
InChI Key GMFQJZUNQAJZDH-UHFFFAOYSA-N
Synonym 2-chloro-4-ethoxyphenyl boronic acid, 2-chloro-4-ethoxypheylboronic acid, 2-chloro-4-ethoxybenzeneboronic acid, boronic acid, 2-chloro-4-ethoxyphenyl, 2-chloro-4-ethoxy phenylboronic acid, 2-chloro-4-ethoxyphenyl boranediol, boronic acid,b-2-chloro-4-ethoxyphenyl, acmc-1ctjv, 4-borono-3-chlorophenethole, 2-chloro-4-ethoxypheylboronicacid
PubChem CID 3539716
IUPAC Name (2-chloro-4-ethoxyphenyl)boronic acid
SMILES CCOC1=CC=C(B(O)O)C(Cl)=C1

Specifications

Melting Point 104°C to 108°C
Quantity 1 g
Formula Weight 200.43
Percent Purity 95%
Chemical Name or Material 2-Chloro-4-ethoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.