Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-fluorobenzaldehyde, 97%

Catalog No. AAA1534506
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1534506 5 g
AAA1534503 1 g
AAA1534514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1534506 Supplier Thermo Scientific Chemicals Supplier No. A1534506
Add to Cart

CAS: 84194-36-5 | C7H4ClFO | 158.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84194-36-5
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.56
MDL Number MFCD00042527
InChI Key KMQWNQKESAHDKD-UHFFFAOYSA-N
Synonym 2-chloro-4-fluoro-benzaldehyde, benzaldehyde, 2-chloro-4-fluoro, pubchem1433, acmc-209pu2, 2-chloro-4-fluorobezaldehyde, ksc448a5h, 2-chloro 4-fluorobenzaldehyde, 4-fluoro-2-chlorobenzaldehyde, 2-chloro 4-fluoro benzaldehyde, timtec-bb sbb003983
PubChem CID 145024
IUPAC Name 2-chloro-4-fluorobenzaldehyde
SMILES FC1=CC=C(C=O)C(Cl)=C1

Specifications

Melting Point 60°C to 63°C
Boiling Point 118°C to 120°C (50 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 3537704
Sensitivity Air Sensitive
Formula Weight 158.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.