Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-fluoronitrobenzene 98.0+%, TCI America™
SDP

Catalog No. C261525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C261525G 25 g
C26155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C261525G Supplier TCI America Supplier No. C261525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2106-50-5
Molecular Formula C6H3ClFNO2
Molecular Weight (g/mol) 175.543
MDL Number MFCD03412200
InChI Key KQOOFMWRLDRDAX-UHFFFAOYSA-N
Synonym 2-chloro-4-fluoronitrobenzene, 1-chloro-5-fluoro-2-nitrobenzene, benzene, 2-chloro-4-fluoro-1-nitro, pubchem4342, acmc-209fh8, 2-choro-4-fluoronitrobenzene, ksc205m1p, 3-chloro-4-nitrofluorobenzene, 4-fluoro-2-chloronitrobenzene, 2-chloro-4-fluoronitrobenzenen
PubChem CID 75017
IUPAC Name 2-chloro-4-fluoro-1-nitrobenzene
SMILES C1=CC(=C(C=C1F)Cl)[N+](=O)[O-]

Specifications

Melting Point 37°C
Color Yellow
Boiling Point 71°C
Quantity 25 g
Formula Weight 175.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-fluoronitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.