Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-fluorotoluene, 98%

Catalog No. AAB2027822
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2027822 100 g
AAB2027814 25 g
AAB2027836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2027822 Supplier Thermo Scientific Chemicals Supplier No. B2027822
Add to Cart

CAS: 452-73-3 | C7H6ClF | 144.573 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-73-3
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.573
MDL Number MFCD00000572
InChI Key CSARJIQZOSVYHA-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorotoluene, benzene, 2-chloro-4-fluoro-1-methyl, 4-fluoro-2-chlorotoluene, 2-chlor-4-fluor-1-methylbenzol, pubchem1605, 2-chloro-4fluorotoluene, acmc-1adyb, 4-fluor-2-chlor-toluol, ksc235q0r, csarjiqzosvyha-uhfffaoysa
PubChem CID 96747
IUPAC Name 2-chloro-4-fluoro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)F)Cl

Specifications

Density 1.198
Boiling Point 154°C to 156°C
Flash Point 50°C (122°F)
Odor Aromatic
Refractive Index 1.499
Quantity 100 g
UN Number UN1993
Beilstein 1931690
Formula Weight 144.58
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-fluorotoluene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 207-209-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.