Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-nitroanisole 98.0+%, TCI America™
SDP

Catalog No. C135225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C135225G 25 g
C13525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C135225G Supplier TCI America Supplier No. C135225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4920-79-0
Molecular Formula C7H6ClNO3
Molecular Weight (g/mol) 187.579
MDL Number MFCD00060681
InChI Key DLJPNXLHWMRQIQ-UHFFFAOYSA-N
Synonym 3-Chloro-4-methoxynitrobenzene
PubChem CID 21027
IUPAC Name 2-chloro-1-methoxy-4-nitrobenzene
SMILES COC1=C(C=C(C=C1)[N+](=O)[O-])Cl

Specifications

Melting Point 95°C
Color Yellow
Quantity 25 g
Formula Weight 187.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-nitroanisole
RTECSNumber BZ8578000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.