Learn More
AdipoGen 2-Chloro-5-ClMethiazole
SDP
Supplier:  AdipoGen CDXC0232G005
Encompass_Preferred
Chemical. CAS 105827-91-6. Formula C4H3Cl2NS. MW 168. Synthetic. Intermediate in the preparation of agrochemicals/pesticides.
Catalog No. 50-256-1419
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.