Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-fluorobenzylamine, 97%, Thermo Scientific™

Catalog No. AAH2681006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2681006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2681006 Supplier Thermo Scientific Chemicals Supplier No. H2681006

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202522-23-4
Molecular Formula C7H7ClFN
Molecular Weight (g/mol) 159.588
MDL Number MFCD06213626
InChI Key IGFBMQSUHFQVFE-UHFFFAOYSA-N
Synonym 2-chloro-5-fluorobenzylamine, 2-chloro-5-fluorobenzyl amine, 2-chloro-5-fluorophenyl methanamine, 2-chloro-5-fluoro-benzylamine, 2-chloro-5-fluorophenyl methylamine, 1-2-chloro-5-fluorophenyl methanamine, pubchem4371, acmc-1cio4, rarechem al bw 2058
PubChem CID 17750669
IUPAC Name (2-chloro-5-fluorophenyl)methanamine
SMILES C1=CC(=C(C=C1F)CN)Cl

Specifications

Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 159.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-fluorobenzylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.