Learn More
Sigma Organic Chemistry 2-Chloro-5-iodopyrimidi | 1G | 32779-38-7 | MFCD06200210
Supplier: Sigma Organic Chemistry SYX003351G

2-Chloro-5-iodopyrimidi , 1G
About this item:
CAS #: 32779-38-7
MDL #: MFCD06200210
Chemical Formula: C4H2ClIN2
Molecular Weight: 240.43
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.