Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-methoxybenzimidazole, 94%

Catalog No. AAL08440MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL08440MD 250 mg
AAL0844003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL08440MD Supplier Thermo Scientific Chemicals Supplier No. L08440MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 15965-54-5 | C8H7ClN2O | 182.607 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15965-54-5
Molecular Formula C8H7ClN2O
Molecular Weight (g/mol) 182.607
MDL Number MFCD00051945
InChI Key FMDGYQOERIOABX-UHFFFAOYSA-N
Synonym 2-chloro-5-methoxybenzimidazole, 2-chloro-5-methoxy-1h-benzo d imidazole, 2-chloro-5-methoxy-1h-benzimidazole, 2-chloro-5-methoxy-1h-1,3-benzodiazole, 2-chloro-5-methoxy-3h-1,3-benzodiazole, acmc-1bs4u, 5-methoxy-2-chlorobenzimidazole, 2-chloro-5-methoxybenzoimidazole, 1h-benzimidazole,2-chloro-6-methoxy, 1h-benzimidazole, 2-chloro-5-methoxy
PubChem CID 519197
IUPAC Name 2-chloro-6-methoxy-1H-benzimidazole
SMILES COC1=CC2=C(C=C1)N=C(N2)Cl

Specifications

Melting Point 165°C to 168°C
Quantity 250 mg
Beilstein 777455
Formula Weight 182.61
Percent Purity 94%
Chemical Name or Material 2-Chloro-5-methoxybenzimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.