Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-5-methoxyphenylboronic acid, 97%

Catalog No. AC430340010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430340010 1 g Glass bottle
AC430340050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC430340010 Supplier Thermo Scientific Chemicals Supplier No. 430340010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 89694-46-2 | C7H8BClO3 | 186.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89694-46-2
Molecular Formula C7H8BClO3
Molecular Weight (g/mol) 186.40
MDL Number MFCD06659858
InChI Key REFXAANPQCJZRY-UHFFFAOYSA-N
Synonym 2-chloro-5-methoxyphenyl boronic acid, 2-chloro-5-methoxyphenylboronicacid, 3-borono-4-chloroanisole, 2-chloro-5-methoxybenzeneboronic acid, 2-chloro-5-methoxy-phenyl boronic acid, boronic acid, 2-chloro-5-methoxyphenyl, pubchem6932, acmc-209r2h, 2-chloro-5-methoxylphenylboronic acid
PubChem CID 17750233
IUPAC Name (2-chloro-5-methoxyphenyl)boronic acid
SMILES COC1=CC=C(Cl)C(=C1)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 111.0°C to 114.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 186.4
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloro-5-methoxyphenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.