Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-nitrophenol 98.0+%, TCI America™
SDP

Catalog No. C207725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C207725G 25 g
C20775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C207725G Supplier TCI America Supplier No. C207725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 619-10-3
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.55
MDL Number MFCD01571825
InChI Key BUMGQSCPTLELLS-UHFFFAOYSA-N
Synonym phenol, 2-chloro-5-nitro, 6-chloro-3-nitrophenol, 2-chloro-5-nitro-phenol, pubchem1475, acmc-209mxh, 2-chloro-5-nitrofenol, 5-nitro-2-chlorophenol, 2-chloro-5-nitro phenol, ksc494c3h, 4-chloro-3-hydroxynitrobenzene
PubChem CID 69264
IUPAC Name 2-chloro-5-nitrophenol
SMILES OC1=CC(=CC=C1Cl)[N+]([O-])=O

Specifications

Melting Point 112°C
Quantity 25 g
Formula Weight 173.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitrophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.