Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-nitrotoluene 98.0+%, TCI America™
SDP

Catalog No. C123325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C123325G 25 g
C1233500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C123325G Supplier TCI America Supplier No. C123325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13290-74-9
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00041430
InChI Key BGDCQZFFNFXYQC-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrotoluene, benzene, 1-chloro-2-methyl-4-nitro, 4-chloro-3-methylnitrobenzene, 1-chloro-2-methyl-4-nitro-benzene, 3-nitro-6-chlorotoluene, 2-methyl-4-nitrochlorobenzene, pubchem2137, 2-chloro-5-nitroluene, acmc-209bqf, 5-nitro-2-chlorotoluene
PubChem CID 83315
IUPAC Name 1-chloro-2-methyl-4-nitrobenzene
SMILES CC1=C(C=CC(=C1)[N+](=O)[O-])Cl

Specifications

Melting Point 44°C
Color Yellow
Boiling Point 96°C
Quantity 25 g
UN Number 3457
Formula Weight 171.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitrotoluene
EINECSNumber (3)-0449
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.